Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tri-Sodium citrate 99.8% for molecular biology DNase-, RNase-, protease-free

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

AC32716-0010 AC32716-0050 AC32716-2500
TS32716-0010EA 61 USD InStock
TS32716-0010 TS32716-0050 TS32716-2500
tri-Sodium citrate 99.8% for molecular biology DNase-, RNase-, protease-free
tri-Sodium citrate hydrate


Formula: C₆H₅Na₃O₇·xH₂O
MW: 258.07 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150032
CAS Number: 114456-61-0
EINECS: 200-675-3
Merck Index: 12,08746
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥99.75 % (on dry substance)
Loss on drying 12 % to 13 %
UV at 260 nm A: ≤0.02
pH 7 to 9 (5 % in water)
Dnase,Rnase,Protease activity None detected
Chloride (Cl) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty