Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel(II) bromide hydrate 98% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

AC21487-1000 AC21487-5000
TS21487-1000EA 117 USD InStock
TS21487-1000 TS21487-5000
Nickel(II) bromide hydrate 98% for analysis
Nickel(II) bromide hydrate


Formula: Br₂Ni.xH₂O
Melting Pt: 963 °C
MDL Number: MFCD00149806
CAS Number: 207569-11-7
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Orange Crystalline powder or needles
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤21 % (K.F.)
pH 5 to 7 (5 % in water)
Sulfate (SO4) ≤50 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤50 ppm
Cobalt (Co) ≤100 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤20 ppm
Magnesium (Mg) ≤20 ppm
Potassium (K) ≤20 ppm
Sodium (Na) ≤500 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty