Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Calcium nitrate tetrahydrate 99+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

42353-0025 AC42353-0250 AC42353-5000
TS42353-0025EA 309 USD InStock
TS42353-0025 TS42353-0250 TS42353-5000
Calcium nitrate tetrahydrate 99+% ACS
Calcium nitrate tetrahydrate


Formula: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Melting Pt: 42
Density: 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
Merck Index: 13,01687
Order Now
Specification Test Results
Appearance White Crystals
Titration Complexometric 99.0 to 103.0 %
pH 5.0 to 7.0 (5 % at 25°C)
Chloride (Cl) ≤50 ppm
Nitrite (NO2) ≤10 ppm
Sulfate (SO4) ≤20 ppm
Barium (Ba) ≤50 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤500 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Strontium (Sr) ≤500 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty