Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloroform 99.8% stabilised for spectroscopy

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

Stabilization: Stabilised with ethanol

40463-5000 404630025
TS40463-5000EA 89 USD InStock
TS40463-5000 TS404630025
Chloroform 99.8% stabilised for spectroscopy
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7
Melting Pt: -63
Density: 1.48 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=96870c784520df552dcde9a71610c883a1c71b50&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
GC ≥99.75 % (Excluding stabilizer)
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % ethanol
Residue after evaporation ≤0.001 %
Acetone and aldehyde ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty