Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dichloromethyl methyl ether 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11347-1000 11347-0100
TS11347-1000EA 155 USD InStock
TS11347-1000 TS11347-0100
Dichloromethyl methyl ether 97%
Dichloromethyl methyl ether


Formula: C₂H₄Cl₂O
MW: 114.96 g/mol
Boiling Pt: 85 °C (760 mmHg)
Density: 1.27 g/cm³
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000828
CAS Number: 4885-02-3
EINECS: 225-498-9
UN: 2924
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4330 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty