Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hexanol 98%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12079-0250 12079-0010 12079-0025 12079-0100 12079-1000
TS12079-0250EA 678 USD InStock
TS12079-0250 TS12079-0010 TS12079-0025 TS12079-0100 TS12079-1000
1-Hexanol 98%, pure
1-Hexanol


Formula: H₃C(CH₂)₄CH₂OH
Boiling Pt: 156…157 °C (760 mmHg)
Melting Pt: –52 °C
Density: 0.8136 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
Merck Index: 13,04715
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=0963e4b94d58210718efb71711e4b68e0fe1dd74&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
Water ≤0.1 %
GC ≥97.5 %
Color scale ≤10 APHA
Acid value ≤0.02 mg KOH/g
Ester value ≤0.3 mg KOH/g
Iodine value ≤0.1 g I2/100 g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty