Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Propanol 99.5% for HPLC

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

38391-0010 38391-0025
TS38391-0010EA 78 USD InStock
TS38391-0010 TS38391-0025
2-Propanol 99.5% for HPLC
2-Propanol


Formula: (CH₃)₂CHOH
Boiling Pt: 81…83 °C (760 mmHg)
Melting Pt: –89.5 °C
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.1 %
GC ≥99.5 %
UV at 240 nm A: ≤0.05
Color scale ≤10 APHA
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0001 meq/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty