Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Propanol 99%, AcroSeal™ Biochemistry, Histology

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

44708-0010 44708-0025
TS44708-0010EA 56 USD InStock
TS44708-0010 TS44708-0025
2-Propanol 99%, AcroSeal™ Biochemistry, Histology
2-Propanol


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.785 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Heavy metals ≤5 ppm
GC ≥98.5 %
UV at 300 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity/alkalinity ≤0.0001 meq/g

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty