Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl perbenzoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17014-0010 17014-2500
TS17014-0010EA 162 USD InStock
TS17014-0010 TS17014-2500
tert-Butyl perbenzoate 98%
tert-Butyl perbenzoate


Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 282
Melting Pt: 8
Density: 1.04 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00008802
CAS Number: 614-45-9
EINECS: 210-382-2
UN: 3103
ADR: 5.2
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5000 (20°C, 589 nm)
Additional info saponifiable chlorine: < 0.035%
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty