Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroquinone monomethyl ether 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12600-0010 12600-0051
TS12600-0010EA 129 USD InStock
TS12600-0010 TS12600-0051
Hydroquinone monomethyl ether 99%
Hydroquinone monomethyl ether


Formula: C₇H₈O₂
Boiling Pt: 243 °C (760 mmHg)
Melting Pt: 56…57 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8
Order Now
Specification Test Results
Appearance White to light yellow Flakes
Infrared spectrum Conforms
Melting point 54 to 57 °C
GC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty