Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-Xylene 99+%, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18086-0010 18086-2500
TS18086-0010EA 180 USD InStock
TS18086-0010 TS18086-2500
m-Xylene 99+%, extra pure
m-Xylene


Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 138
Melting Pt: -48
Density: 0.864 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3 III
Merck Index: 12,10214
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %
Residue after evaporation ≤0.005 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty