Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexane 99.5% for analysis

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

17681-0010 17681-0025 17681-0250 176810050 176815000
TS17681-0010EA 76 USD InStock
TS17681-0010 TS17681-0025 TS17681-0250 TS176810050 TS176815000
Cyclohexane 99.5% for analysis
Cyclohexane


Formula: C₆H₁₂
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 6.5 °C
Density: 0.77 g/cm³
Flash Pt: –18 °C
Storage Temperature: Ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=70ea12051c5d4a25e80a997bdd0c07cd94520e81&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.05 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Substance darkened by H2SO4 ≤10 APHA
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Potassium (K) ≤0.2 ppm
Sodium (Na) ≤1 ppm
Zinc (Zn) ≤0.1 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty