Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium chloride 99+% ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

42429-0010 42429-0050 42429-0251 AC42429-0250 AC42429-5000
TS42429-0010EA 114 USD InStock
TS42429-0010 TS42429-0050 TS42429-0251 TS42429-0250 TS42429-5000
Sodium chloride 99+% ACS
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599
Order Now
Specification Test Results
Appearance White Crystals
Titration Argentometric ≥99.0 %
Loss on drying ≤0.5 % (1 g, 105°C, 2 hrs)
pH 5.0 to 9.0 (5 % solution at 25.0°C)
Bromide (Br) ≤0.01 %
Iodide (I) ≤0.002 %
Chlorate and Nitrate (as NO3) ≤0.003 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.004 %
Barium (Ba) Passes test
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤2 ppm
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty