Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sulfuric acid standard solution 0.5 mol/l (1 N) in water

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12424-0010 12424-0025
TS12424-0010EA 54 USD InStock
TS12424-0010 TS12424-0025
Sulfuric acid standard solution 0.5 mol/l (1 N) in water
Sulfuric acid


Formula: H₂SO₄
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty