Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Antimony(III) chloride 99.5%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20168-5000 AC20168-0020
TS20168-5000EA 145 USD InStock
TS20168-5000 TS20168-0020
Antimony(III) chloride 99.5%
Antimony(III) chloride


Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5
Melting Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00011212
CAS Number: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8 II
Merck Index: 13,00713
Order Now
Specification Test Results
Appearance Colorless to white to off-white Adhering crystals and/or chunks (May have a pink hue)
Titration Iodimetric ≥99.4 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty