Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Oxalyl dichloride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

12961-0250 12961-1000 12961-0010 12961-0025
TS12961-0250EA 55 USD InStock
TS12961-0250 TS12961-1000 TS12961-0010 TS12961-0025
Oxalyl dichloride 98%
Oxalyl dichloride


Formula: C₂Cl₂O₂
MW: 126.93 g/mol
Boiling Pt: 63
Melting Pt: -12
Density: 1.478 g/cm³
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000704
CAS Number: 79-37-8
EINECS: 201-200-2
UN: 3130
ADR: 4.3 I
Merck Index: 12,07046
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4285 to 1.4305 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty