Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

34844-0010 34844-5000 348440025 348441000
TS34844-0010EA 179 USD InStock
TS34844-0010 TS34844-5000 TS348440025 TS348441000
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal™
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=d0727d3220885f6adab5f099d9fd3b4de38d5710&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.70 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty