Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl methanesulphonate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20526-1000 20526-0100 20526-0250 20526-5000
TS20526-1000EA 412 USD InStock
TS20526-1000 TS20526-0100 TS20526-0250 TS20526-5000
Ethyl methanesulphonate 99%
Ethyl methanesulphonate


Formula: C₃H₈O₃S
MW: 124.16 g/mol
Boiling Pt: 213
Melting Pt: –96 °C
Density: 0.92 g/cm³
Flash Pt: 6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007559
CAS Number: 62-50-0
EINECS: 209-734-8
UN: 2333
ADR: 3 II
Merck Index: 13,03859
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty