Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyric anhydride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

10812-5000 10812-0025 10812-1000
TS10812-5000EA 64 USD InStock
TS10812-5000 TS10812-0025 TS10812-1000
Butyric anhydride 98%
Butyric anhydride


Formula: C₈H₁₄O₃
MW: 158.2 g/mol
Boiling Pt: 198
Melting Pt: -73
Density: 0.963 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00009389
CAS Number: 106-31-0
EINECS: 203-383-4
UN: 2739
ADR: 8 III
Merck Index: 13,01594
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4140 (20°C, 589 nm)
GC ≥97.5 %
UV at 260 nm A: ≤0.15
Appearance of solution faint yellow
NMR (1H-NMR) Chondroitin sulfate (over-sulfated): Absent
Protein ≤0.5 % (dried substance)
pH 5.5 to 8.0 (0.1g in 10ml water)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty