Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dihydroxynaphthalene 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16334-0050 16334-0010 16334-0250
TS16334-0050EA 156 USD InStock
TS16334-0050 TS16334-0010 TS16334-0250
1,3-Dihydroxynaphthalene 99+%
1,3-Dihydroxynaphthalene


Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 386
Melting Pt: 124
Storage Temperature: Ambient
MDL Number: MFCD00003965
CAS Number: 132-86-5
EINECS: 205-079-7
Merck Index: 12,06483
Order Now
Specification Test Results
Appearance Pink-orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
Titration with NaOH ≥99.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty