Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium bis(trimethylsilyl)amide 95+%, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21596-0250 AC21596-0050 AC21596-1000
TS21596-0250EA 150 USD InStock
TS21596-0250 TS21596-0050 TS21596-1000
Sodium bis(trimethylsilyl)amide 95+%, pure
Sodium bis(trimethylsilyl)amide


Formula: C₆H₁₈NNaSi₂
MW: 183.38 g/mol
Melting Pt: 171…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009835
CAS Number: 1070-89-9
EINECS: 213-983-8
UN: 3096
ADR: 8 I
Order Now
Specification Test Results
Appearance White to light yellow to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 95.0 to 105.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty