Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium methoxide 30% (w/w) 5.4 M in methanol, pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16860-0025 AC16860-0010 AC16860-0100 AC16860-0250 AC16860-0251
TS16860-0025EA 142 USD InStock
TS16860-0025 TS16860-0010 TS16860-0100 TS16860-0250 TS16860-0251
Sodium methoxide 30% (w/w) 5.4 M in methanol, pure
Sodium methoxide


Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 127
Density: 0.97 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1289
ADR: 3 III
Order Now
Specification Test Results
Appearance Colorless to light yellow Solution
Strength 5.3 to 5.6 M
Titration Argentometric ≥94.0 to ≤106.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty