Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Propionamide 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

16267-1000 16267-0250
TS16267-1000EA 135 USD InStock
TS16267-1000 TS16267-0250
Propionamide 97%
Propionamide


Formula: C₃H₇NO
Boiling Pt: 197…200 °C (760 mmHg)
Melting Pt: –49 °C
Density: 0.84 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00008039
CAS Number: 79-05-0
EINECS: 203-918-1
UN: 3082
ADR: 9,III
Merck Index: 12,08009
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4525 to 1.4545 (20°C, 589 nm)
GC ≥96.0 %
Titration with NaOH ≥96.0 to ≤104.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty