Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl sulfate 99+%, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

43083-1000 AC43083-0010
TS43083-1000EA 43 USD InStock
TS43083-1000 TS43083-0010
Dimethyl sulfate 99+%, AcroSeal™
Dimethyl sulphate


Formula: C₂H₆O₄S
Boiling Pt: 188 °C (760 mmHg)
Melting Pt: –32 °C
Density: 1.33 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008416
CAS Number: 77-78-1
UN: 1595
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20°C, 589 nm)
Acidity ≤0.2 % (as Sulfuric acid)
Titration 99.0 to 101.0 % (Hydrolysis)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty