Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-Platin 99.99% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

19376-0010 19376-0050 AC19376-2500
TS19376-0010EA 260 USD InStock
TS19376-0010 TS19376-0050 TS19376-2500
cis-Platin 99.99% (trace metals basis)
cis-Platin


Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Melting Pt: 277 °C
Storage Temperature: Ambient
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1 II
Merck Index: 13,02341
Order Now
Specification Test Results
Appearance Yellow to orange Powder or crystals
Infrared spectrum Conforms
Assay 64.0 to 66.0 % (Pt)
Total trace metal impurities ≤0.01 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty