Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Yttrium (III) nitrate hexahydrate 99.9% (trace metals basis)

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

21826-5000 AC21826-0250 AC21826-1000
TS21826-5000EA 415 USD InStock
TS21826-5000 TS21826-0250 TS21826-1000
Yttrium (III) nitrate hexahydrate 99.9% (trace metals basis)
Yttrium (III) nitrate hexahydrate


Formula: N₃O₉Y·6H₂O
MW: 383.01 g/mol
Melting Pt: 51.8
Storage Temperature: Ambient
MDL Number: MFCD00149940
CAS Number: 13494-98-9
EINECS: 233-802-6
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White Adhering crystalline powder or crystals
Total trace metal impurities ≤0.1 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty