Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,10-Phenanthroline monohydrate ACS

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

41712-1000 41712-0010 41712-0050 41712-0250
TS41712-1000EA 652 USD InStock
TS41712-1000 TS41712-0010 TS41712-0050 TS41712-0250
1,10-Phenanthroline monohydrate ACS
1,10-Phenanthroline monohydrate


Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00149973
CAS Number: 5144-89-8
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,07291
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba346b9f67fdda9afd4d48b893e6318564d604c&vid=aba346b9f67fdda9afd4d48b893e6318564d604c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Crystalline powder or needles (May discolor to cream on storage)
Infrared spectrum Conforms
Suitability (for Fe determination) Passes test
UV at 450 nm A: ≤0.01
Color scale ≤20 APHA

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty