Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl trifluoroacetate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11902-1000 11902-0025 11902-0250 11902-5000
TS11902-1000EA 140 USD InStock
TS11902-1000 TS11902-0025 TS11902-0250 TS11902-5000
Ethyl trifluoroacetate 99%
Ethyl trifluoroacetate


Formula: C₄H₅F₃O₂
MW: 142.08 g/mol
Boiling Pt: 61
Melting Pt: -78
Density: 0.86 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 208-760-7
UN: 1123
ADR: 3 II
Merck Index: 13,01536
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3880 (20°C, 589 nm)
GC ≥98.5 %
Residue after evaporation ≤0.002 %
Titration with NaOH ≥88.0 %
Dilution test passes test
Chloride (Cl) ≤10 ppm
Sulfite (SO3) passes test
Sulfate (SO4) ≤20 ppm
Ammonium (NH4) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Free acid (CH3COOH) ≤0.4 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty