Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrabutylammonium fluoride trihydrate 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22108-2500 AC22108-0100 AC22108-0500
TS22108-2500EA 665 USD InStock
TS22108-2500 TS22108-0100 TS22108-0500
Tetrabutylammonium fluoride trihydrate 99%
Tetrabutylammonium fluoride trihydrate


Formula: C₁₆H₃₆FN·3H₂O
MW: 315.51 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 1759
ADR: 8 III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty