Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium cerium(IV) sulphate dihydrate 99+%, extra pure

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

20631-1000 AC20631-0050 AC20631-5000
TS20631-1000EA 117 USD InStock
TS20631-1000 TS20631-0050 TS20631-5000
Ammonium cerium(IV) sulphate dihydrate 99+%, extra pure
Ammonium cerium(IV) sulphate dihydrate


Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
Melting Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00149417
CAS Number: 10378-47-9
EINECS: 231-567-4
Order Now
Specification Test Results
Appearance Yellow to orange Crystalline powder
Titration with Na2S2O3 ≥99.0 %
Chloride (Cl) ≤100 ppm
Phosphate (PO4) ≤100 ppm
Iron (Fe) ≤50 ppm
Insoluble matter ≤0.02 % (in dilute H2SO4)

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty