Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (±)-lactate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

41448-0050 41448-0250
TS41448-0050EA 87 USD InStock
TS41448-0050 TS41448-0250
Methyl (±)-lactate 98%
Methyl (±)-lactate


Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 35
Melting Pt: –32 °C
Density: 0.69 g/cm³
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00066367
CAS Number: 547-64-8
EINECS: 207-962-2
UN: 1144
ADR: 3 I
Merck Index: 12,06167
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3920 to 1.3940 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (neat)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty