Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hexacarbonylmolybdenum 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

19039-0500 19039-2500 AC19039-0100
TS19039-0500EA 483 USD InStock
TS19039-0500 TS19039-2500 TS19039-0100
Hexacarbonylmolybdenum 98%
Hexacarbonylmolybdenum


Formula: C₆MoO₆
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: 150 °C (dec.)
Density: 1.96 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003466
CAS Number: 13939-06-5
EINECS: 237-713-3
UN: 3466
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty