Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-2-methyl-1,3-propanediol 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

10405-1000 10405-5000
TS10405-1000EA 81 USD InStock
TS10405-1000 TS10405-5000
2-Amino-2-methyl-1,3-propanediol 99%
2-Amino-2-methyl-1,3-propanediol


Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 272
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8 III
Merck Index: 13,00447
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point ≥100 °C
Water ≤0.5 %
Titration with HClO4 ≥98.5 % (on anhydrous substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty