Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dicyclohexylcarbodiimide 99%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

11390-1000 11390-0010 11390-0050 11390-0250 11390-2500
TS11390-1000EA 86 USD InStock
TS11390-1000 TS11390-0010 TS11390-0050 TS11390-0250 TS11390-2500
Dicyclohexylcarbodiimide 99%
Dicyclohexylcarbodiimide


Formula: C₁₃H₂₂N₂
MW: 206.33 g/mol
Boiling Pt: 122…124 °C (6 mmHg)
Melting Pt: 33…35 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00011659
CAS Number: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1 II
Merck Index: 13,03123
Order Now
Specification Test Results
Appearance White to light yellow Crystalline mass or waxy solid
Infrared spectrum Conforms
Melting point 33 to 35 °C
GC ≥98.5 %
Appearance of solution (10% in acetone) clear colorless to yellowish
Color scale ≤25 APHA (ASTM D 1209)
Bromine number ≤5 g Br/100 g (ASTM D 1159)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty