Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methylmagnesium bromide 3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39112-8000 AC39112-1000
TS39112-8000EA 397 USD InStock
TS39112-8000 TS39112-1000
Methylmagnesium bromide 3.2 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Methylmagnesium bromide


Formula: CH₃BrMg
MW: 119.24 g/mol
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000041
CAS Number: 75-16-1
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Dark brown Solution (May develop some turbidity or precipitate)
Titration Grignard 34 to 36 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty