Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiocarbonyl chloride 85%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

15164-1000 15164-0025 15164-5000
TS15164-1000EA 256 USD InStock
TS15164-1000 TS15164-0025 TS15164-5000
Thiocarbonyl chloride 85%
Thiocarbonyl chloride


Formula: CCl₂S
Boiling Pt: 73 °C (760 mmHg)
Density: 1.508 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004918
CAS Number: 463-71-8
EINECS: 207-341-6
UN: 2474
ADR: 6.1,
Order Now
Specification Test Results
Appearance Orange-red to red Liquid
GC ≥80 %
Melting point 164 to 169 °C
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty