Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,8-Diazabicyclo[5.4.0]undec-7-ene 98+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

16061-1000 16061-5000 16061-0025 16061-0250
TS16061-1000EA 94 USD InStock
TS16061-1000 TS16061-5000 TS16061-0025 TS16061-0250
1,8-Diazabicyclo[5.4.0]undec-7-ene 98+%
1,8-Diazabicyclo[5.4.0]undec-7-ene


Formula: C₉H₁₆N₂
MW: 152.24 g/mol
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: –70 °C
Density: 1.019 g/cm³
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006930
CAS Number: 6674-22-2
EINECS: 229-713-7
UN: 2922
ADR: 8 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5230 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty