Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl hydroperoxide 70% in water

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

18034-5000 18034-0025 18034-0050 18034-0100 18034-2500
TS18034-5000EA 58 USD InStock
TS18034-5000 TS18034-0025 TS18034-0050 TS18034-0100 TS18034-2500
tert-Butyl hydroperoxide 70% in water
tert-Butyl hydroperoxide


Formula: C₄H₁₀O₂
MW: 90.12
Boiling Pt: 96 °C (dec.)
Melting Pt: –3 °C
Density: 0.937
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002130
CAS Number: 75-91-2
EINECS: 200-915-7
UN: 3109
ADR: 5.2
Merck Index: 14,1570
Order Now
Specification Test Results
Appearance Clear solution
Titration Iodometric 68.5 to 71.0 %
Color scale ≤20 APHA
Titration with HClO4 ≥97.5 % (on dry substance)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty