Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzeneseleninic anhydride 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

19971-0250 AC19971-0050
TS19971-0250EA 420 USD InStock
TS19971-0250 TS19971-0050
Benzeneseleninic anhydride 98%
Benzeneseleninic anhydride


Formula: C₁₂H₁₀O₃Se₂
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00001991
CAS Number: 17697-12-0
EINECS: 241-701-3
UN: 3283
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to yellow to light beige Crystalline powder or needles
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty