Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Silicone oil temperature_high

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

17466-0025 17466-5000 AC17466-1000
TS17466-0025EA 994 USD InStock
TS17466-0025 TS17466-5000 TS17466-1000
Silicone oil temperature_high
Silicone oil


Density: 1.07 g/cm³
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Order Now
Specification Test Results
Appearance Clear colorless to straw yellow Oily liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.5040 (20°C, 589 nm)
Viscosity 100 to 150 cSt (at 25°C)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty