Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Mercaptoethanesulphonic acid sodium salt (mesna) 98% analytical standard

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

44315-0250 44315-0050 44315-0100
TS44315-0250EA 214 USD InStock
TS44315-0250 TS44315-0050 TS44315-0100
2-Mercaptoethanesulphonic acid sodium salt (mesna) 98% analytical standard
2-Mercaptoethanesulphonic acid sodium salt (mesna)


Formula: C₂H₅NaO₃S₂
Storage Temperature: Ambient
MDL Number: MFCD00007535
CAS Number: 19767-45-4
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 to 102 % (On dried substance)
Loss on drying ≤2 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty