Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-Ethylhydroxylamine hydrochloride 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

40961-0100 AC40961-0250
TS40961-0100EA 270 USD InStock
TS40961-0100 TS40961-0250
O-Ethylhydroxylamine hydrochloride 99+%
O-Ethylhydroxylamine hydrochloride


Formula: C₂H₈ClNO
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00012956
CAS Number: 3332-29-4
EINECS: 222-060-9
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
HPLC ≥98.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty