Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vinylphosphonic acid 90%, Technical Grade

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

30059-2500 30059-0100 30059-0500
TS30059-2500EA 311 USD InStock
TS30059-2500 TS30059-0100 TS30059-0500
Vinylphosphonic acid 90%, Technical Grade
Vinylphosphonic acid


Formula: C₂H₅O₃P
MW: 108.0331 g/mol
Melting Pt: 36 °C
Density: 1.37 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00043866
CAS Number: 1746-03-8
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Titration Acid ≥88 %
Water ≤0.5 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty