Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2':6',2''-Terpyridine 96%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

21563-0010 21563-0050 21563-2500
TS21563-0010EA 152 USD InStock
TS21563-0010 TS21563-0050 TS21563-2500
2,2':6',2''-Terpyridine 96%
2,2':6',2''-Terpyridine


Formula: C₁₅H₁₁N₃
Boiling Pt: 370 °C
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006213
CAS Number: 1148-79-4
UN: 2811
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Cream to yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 95 °C
GC ≥95 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty