Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethylzinc 1.2 mol/l in toluene, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

37724-1000 377248000
TS37724-1000EA 280 USD InStock
TS37724-1000 TS377248000
Dimethylzinc 1.2 mol/l in toluene, AcroSeal™
Dimethylzinc


Formula: C₂H₆Zn
Storage Temperature: Ambient
MDL Number: MFCD00014854
CAS Number: 544-97-8
EINECS: 208-884-1
UN: 3399
Order Now
Specification Test Results
Appearance Clear colorless Solution
Concentration 11.5 to 12.5 wt% (On Zn basis)

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty