Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Furaldehyde 98% stabilized

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

39326-0250 39326-0010 39326-0050
TS39326-0250EA 323 USD InStock
TS39326-0250 TS39326-0010 TS39326-0050
3-Furaldehyde 98% stabilized
3-Furaldehyde


Formula: C₅H₄O₂
Boiling Pt: 145 °C (760 mmHg)
Density: 1.15 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010424
CAS Number: 498-60-2
UN: 1199
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4950 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.05 % BHT

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty