Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cholesterol benzoate 98%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

11020-1000 11020-0250
TS11020-1000EA 199 USD InStock
TS11020-1000 TS11020-0250
Cholesterol benzoate 98%
Cholesterol benzoate


Formula: C₃₄H₅₀O₂
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003635
CAS Number: 604-32-0
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration RCOCl ≥97.5 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty