Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glyoxalbis(2-hydroxyanil) 97%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

12016-0100 12016-0500
TS12016-0100EA 94 USD InStock
TS12016-0100 TS12016-0500
Glyoxalbis(2-hydroxyanil) 97%
Glyoxalbis(2-hydroxyanil)


Formula: C₁₄H₁₂N₂O₂
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00002183
CAS Number: 1149-16-2
Order Now
Specification Test Results
Appearance Beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Sulfated ash ≤0.1 % (800°C)
Solubility (0.5 % in methanol) Clear
HPLC ≥96.0 %
Suitability (for Ca determination) Passes test
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤200 ppm

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty