Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Vinylmagnesium bromide 0.7 M in THF, AcroSeal™

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

20939-1000 AC20939-8000
TS20939-1000EA 68 USD InStock
TS20939-1000 TS20939-8000
Vinylmagnesium bromide 0.7 M in THF, AcroSeal™
Vinylmagnesium bromide


Formula: C₂H₃BrMg
MW: 131.26 g/mol
Density: 0.98 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000042
CAS Number: 1826-67-1
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Clear yellow to orange to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 0.70 to 0.90 M

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty