Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-4-nitrophenyl sulfate 99+%

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Danger

22749-0050 22749-0010
TS22749-0050EA 347 USD InStock
TS22749-0050 TS22749-0010
Potassium-4-nitrophenyl sulfate 99+%
Potassium-4-nitrophenyl sulfate


Formula: C₆H₄KNO₆S
MW: 257.257 g/mol
Boiling Pt: 37 °C (760 mmHg)
Melting Pt: –151 °C
Density: 0.65 g/cm³
Flash Pt: –27 °C
Storage Temperature: Ambient
MDL Number: MFCD00007466
CAS Number: 6217-68-1
EINECS: 210-988-7
UN: 3295
ADR: 3 II
Merck Index: 12,07263
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3810 to 1.3830 (20°C, 589 nm)
GC ≥95.0 %

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty